missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, MilliporeSigma™

Catalog No. M8017912500
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M8017912500 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M8017912500 Supplier Technidata Supplier No. 8.01791.2500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For synthesis

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.556
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
Synonym monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
PubChem CID 7964
ChEBI CHEBI:28097
IUPAC Name chlorobenzene
SMILES C1=CC=C(C=C1)Cl

Specifications

pH Neutral (at 20°C)
Melting Point -45°C
Color Colorless
Density 1.11g/cm3 at 20°C
Quantity 2.5 L
Flash Point 27°C (80.6°F)
Odor Benzene-like
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.52
Vapor Pressure 12 hPa (20°C)
UN Number 1134
Boiling Point 132°C
Solubility Information Soluble in water: 0.4g/L at 20°C
Formula Weight 112.56
Chemical Name or Material Chlorobenzene
Show More Show Less
Hazard Category H226: Flammable liquid and vapour.
Hazard Statement H315 Causes skin irritation.
Precautionary Statement H332: Harmful if inhaled.
Recommended Storage 15° to 25°C (59° to 77°F); Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition.