missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, ACS Reagent (Monochlorobenzene), Reagents

Catalog No. 01337325
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337325 4 L
01337324 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337325 Supplier Labchem Inc Supplier No. C21751004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chlorobenzene, ACS Reagent (Monochlorobenzene), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.56
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
Synonym Monochlorobenzene, Phenyl chloride, Benzene chloride
IUPAC Name chlorobenzene
SMILES ClC1=CC=CC=C1

Specifications

Melting Point -45°C
Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.107
Boiling Point 132°C
Grade ACS
Chemical Name or Material Chlorobenzene, ACS Reagent (Monochlorobenzene)