Learn More
Chlorobenzene, ACS, 99.5%
Description
Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -45°C |
| Color | Colorless |
| Density | 1.106 g/mL |
| Quantity | 4 L |
| Flash Point | 28°C (82°F) |
| Odor | Aromatic |
| Physical Form | Liquid |
| Refractive Index | 1.524 |
| UN Number | UN1134 |
| Beilstein | 605632 |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H332 |
| Hazard Statement | GHS H Statement H226-H332 Flammable liquid and vapor. Harmful if inhaled. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZENE |
| EINECSNumber | 203-628-5 |
| RTECSNumber | CZ0175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only