Learn More
Chlorobenzene, 99.8%, Extra Dry, AcroSeal™
CAS: 108-90-7 | C6H5Cl | 112.556 g/mol
Supplier: Thermo Scientific Chemicals 396970010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Chlorobenzene | |
| Extra Dry | |
| Colorless | |
| 131.0°C | |
| 23°C | |
| Liquid | |
| 0.001% max. | |
| 1.1075 | |
| C6H5Cl | |
| 0.8 mPa.s (20°C) | |
| 0.005% Max. (Coulometric) | |
| monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene | |
| MVPPADPHJFYWMZ-UHFFFAOYSA-N | |
| chlorobenzene | |
| 7964 | |
| 112.56 | |
| Extra Dry |
| -45.0°C | |
| 108-90-7 | |
| 1.1075g/mL | |
| 1 L | |
| AcroSeal™ Glass Bottle | |
| 1.5230 to 1.5250 | |
| 99.8% | |
| C6H5Cl | |
| MFCD00000530 | |
| 1134 | |
| 15, 2122 | |
| Solubility in water: 0.4g/L (20°C). Other solubilities: miscible with most common organic solvents | |
| C1=CC=C(C=C1)Cl | |
| 112.556 | |
| CHEBI:28097 | |
| 99.8% |
Chemical Identifiers
| 108-90-7 | |
| 112.556 | |
| MVPPADPHJFYWMZ-UHFFFAOYSA-N | |
| 7964 | |
| chlorobenzene |
| C6H5Cl | |
| MFCD00000530 | |
| monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene | |
| CHEBI:28097 | |
| C1=CC=C(C=C1)Cl |
Safety and Handling
GHS P Statement
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
GHS P Statement
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge
Warning
RUO – Research Use Only