missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, ≥99.5%, ACS reagent

Catalog No. AC404490025
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC404490025 2.5 kg Glass Bottle
AC404490010 1 kg EcoSafPak™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC404490025 Supplier Thermo Scientific Chemicals Supplier No. 404490025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-90-7 | C6H5Cl | 112.556 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.556
MDL Number MFCD00000530
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
Synonym monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
PubChem CID 7964
ChEBI CHEBI:28097
IUPAC Name chlorobenzene
SMILES C1=CC=C(C=C1)Cl

Specifications

Melting Point -45.0°C
Density 1.1075g/mL
Boiling Point 131.0°C
Quantity 2.5 kg
Flash Point 23°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.5230 to 1.5250
Assay Percent Range 99.6%
Residue after Evaporation 0.02% max.
Specific Gravity 1.1075
Titratable Acid 0.004 meq/g max.
Linear Formula C6H5Cl
Vapor Pressure Vapor Pressure: 12mbar at 20°C
UN Number 1134
Viscosity 0.8 mPa.s (20°C)
Beilstein 05, 199
Merck Index 15, 2122
Solubility Information Solubility in water: 0.4g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 112.56
Percent Purity 99.6%
Grade ACS Reagent
Chemical Name or Material Chlorobenzene, For analysis ACS
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

RUO – Research Use Only