missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetonitrile, 98+%

Catalog No. AAA1282536
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1282536 500 g
AAA12825A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1282536 Supplier Thermo Scientific Chemicals Supplier No. A1282536
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 107-14-2 | C2H2ClN | 75.495 g/mol

Chloroacetonitrile is used in the electrochemical synthesis of cyanoacetic acid with carbon dioxide. It is involved in phase-transfer-catalyzed Darzen's condensation reaction with cyclohexanone. It is also used as an eluent additive in thermospray liquid chromatography/mass spectrometry. Further, it is used to prepare polysubstituted pyrido[1,2-a]benzimidazole by reacting with other reactant such as malononitrile, aromatic aldehyde and pyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetonitrile is used in the electrochemical synthesis of cyanoacetic acid with carbon dioxide. It is involved in phase-transfer-catalyzed Darzen′s condensation reaction with cyclohexanone. It is also used as an eluent additive in thermospray liquid chromatography/mass spectrometry. Further, it is used to prepare polysubstituted pyrido[1,2-a]benzimidazole by reacting with other reactant such as malononitrile, aromatic aldehyde and pyridine.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and acids.

Chemical Identifiers

CAS 107-14-2
Molecular Formula C2H2ClN
Molecular Weight (g/mol) 75.495
MDL Number MFCD00001885
InChI Key RENMDAKOXSCIGH-UHFFFAOYSA-N
Synonym chloroacetonitrile, chloracetonitrile, acetonitrile, chloro, chloromethyl cyanide, monochloroacetonitrile, chloroethanenitrile, monochloromethyl cyanide, usaf kf-5, alpha-chloroacetonitrile, acetonitrile, 2-chloro
PubChem CID 7856
ChEBI CHEBI:82416
IUPAC Name 2-chloroacetonitrile
SMILES C(C#N)Cl

Specifications

Density 1.195
Boiling Point 124°C to 126°C
Flash Point 56°C (132°F)
Refractive Index 1.423
Linear Formula ClCH2CN
Quantity 500 g
UN Number UN2668
Beilstein 506028
Solubility Information Immiscible with water.
Formula Weight 75.5
Percent Purity ≥98%
Chemical Name or Material Chloroacetonitrile
Show More Show Less
Hazard Category H226-H301+H331-H310-H319
Hazard Statement GHS H Statement
H301-H311-H331-H226
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P262-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: CHLOROACETONITRILE
EINECSNumber 203-467-0
RTECSNumber AL8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only