missing translation for 'onlineSavingsMsg'
Learn More

chloroacetic anhydride, 97%

Catalog No. AC404420250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC404420250 25 g Glass bottle
AC404421000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC404420250 Supplier Thermo Scientific Chemicals Supplier No. 404420250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 541-88-8 | C4H4Cl2O3 | 170.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 541-88-8
Molecular Formula C4H4Cl2O3
Molecular Weight (g/mol) 170.97
MDL Number MFCD00000929
InChI Key PNVPNXKRAUBJGW-UHFFFAOYSA-N
Synonym chloroacetic anhydride, chloracetic anhydride, monochloroacetic acid anhydride, 2-chloroacetic anhydride, chloroacetic acid anhydride, chloroacetyl anhydride, acetic acid, chloro-, anhydride, unii-8q7txh7r5f, monochloroacetic anhydride, 8q7txh7r5f
PubChem CID 10946
IUPAC Name (2-chloroacetyl) 2-chloroacetate
SMILES ClCC(=O)OC(=O)CCl

Specifications

Melting Point 51°C to 59°C
Color Brown
Boiling Point 203°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (ClCH2CO)2O
Quantity 25 g
Beilstein 02, 199
Fieser 01,129
Merck Index 15, 2113
Solubility Information Solubility in water: decomposes. Other solubilities: freely soluble in ether and chloroform,slightly soluble in benzene,practically insoluble in cold ligroin
Formula Weight 170.98
Percent Purity 97%
Physical Form Crystals, Chunks or Low Melting Mass
Chemical Name or Material chloroacetic anhydride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 208-794-2

RUO – Research Use Only