missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetic Acid 99.0+%, TCI America™

Catalog No. C212325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C212325G 25 g
C2123500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C212325G Supplier TCI America Supplier No. C212325G

Chemical Identifiers

CAS 79-11-8
Molecular Formula C2H3ClO2
Molecular Weight (g/mol) 94.494
MDL Number MFCD00002683
InChI Key FOCAUTSVDIKZOP-UHFFFAOYSA-N
Synonym chloroacetic acid, monochloroacetic acid, chloracetic acid, acetic acid, chloro, acide chloracetique, chloroethanoic acid, monochloroethanoic acid, monochloracetic acid, monochloorazijnzuur, monochloressigsaeure
PubChem CID 300
ChEBI CHEBI:27869
IUPAC Name 2-chloroacetic acid
SMILES C(C(=O)O)Cl

Specifications

Melting Point 65°C
Color White-Yellow
pH <1 (800g/L H2O soln. 20°C)
Boiling Point 189°C
Quantity 25 g
UN Number 1751
Formula Weight 94.49
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Chloroacetic Acid
EINECSNumber (2)-1145
RTECSNumber AF8575000
TSCA Yes