missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetic acid sodium salt, 98%

Catalog No. AC404410025
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC404410025 2.5 kg Glass Bottle
AC404415000 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC404410025 Supplier Thermo Scientific Chemicals Supplier No. 404410025

missing translation for 'validMainframeMsg'

CAS: 3926-62-3 | C2H2ClNaO2 | 116.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3926-62-3
Molecular Formula C2H2ClNaO2
Molecular Weight (g/mol) 116.48
MDL Number MFCD00002684
InChI Key FDRCDNZGSXJAFP-UHFFFAOYSA-M
Synonym sodium chloroacetate, chloroacetic acid sodium salt, monoxone, acetic acid, chloro-, sodium salt, dow defoliant, sodium monochloroacetate, smca, sodium monochloracetate, caswell no. 755a, chloroctan sodny czech
PubChem CID 23665759
IUPAC Name sodium;2-chloroacetate
SMILES C(C(=O)[O-])Cl.[Na+]

Specifications

Melting Point 199.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula CH2ClCOONa
Quantity 2.5 kg
Merck Index 15, 2112
Solubility Information Solubility in water: soluble in water: 440g/L (20°C).. Other solubilities: slightly soluble in methanol., insoluble in acetone, benzene, ether, ccl4.
Formula Weight 116.48
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Chloroacetic acid sodium salt, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
If skin irritation occurs: Get medical advice/attention.
EINECSNumber 223-498-3
RTECSNumber AG1400000
TSCA TSCA

RUO – Research Use Only