missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetic acid, 99+%, ACS reagent

Catalog No. AC448115000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC448115000 500 g Plastic bottle
AC448111000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC448115000 Supplier Thermo Scientific Chemicals Supplier No. 448115000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79-11-8 | C2H3ClO2 | 94.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79-11-8
Molecular Formula C2H3ClO2
Molecular Weight (g/mol) 94.5
InChI Key FOCAUTSVDIKZOP-UHFFFAOYSA-N
Synonym chloroacetic acid, monochloroacetic acid, chloracetic acid, acetic acid, chloro, acide chloracetique, chloroethanoic acid, monochloroethanoic acid, monochloracetic acid, monochloorazijnzuur, monochloressigsaeure
PubChem CID 300
ChEBI CHEBI:27869
IUPAC Name 2-chloroacetic acid
SMILES C(C(=O)O)Cl

Specifications

Melting Point 61.0°C to 63.0°C
Density 1.5800g/mL
Boiling Point 189.0°C
Flash Point 126°C
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula ClCH2CO2H
Quantity 500 g
Merck Index 15,2112
Specific Gravity 1.58
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, chloroform, ether
Viscosity 1.29 Pas (100°C)
Formula Weight 94.5
Percent Purity 99+%
Grade ACS Reagent
Insoluble Matter 0.01% max
Chemical Name or Material Chloroacetic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
May cause respiratory irritation.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rins
EINECSNumber 201-178-4

RUO – Research Use Only