missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroacetamidine hydrochloride, 96%

Catalog No. AAA1483514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1483514 25 g
AAA1483506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1483514 Supplier Thermo Scientific Chemicals Supplier No. A1483514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10300-69-3 | C2H6Cl2N2 | 128.98 g/mol

Chloroacetamidine hydrochloride is used as a general pharmacophore in delineating characteristics of the amidinotransferase superfamily.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetamidine hydrochloride is used as a general pharmacophore in delineating characteristics of the amidinotransferase superfamily.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 10300-69-3
Molecular Formula C2H6Cl2N2
Molecular Weight (g/mol) 128.98
MDL Number MFCD00053013
InChI Key GUPOZVHRTJYZCX-UHFFFAOYSA-N
Synonym chloroacetamidine hydrochloride, 2-chloroacetimidamide hydrochloride, 2-chloroethanimidamide hydrochloride, 2-chloroacetamidine hydrochloride, 2-chloroacetamidine hcl, 2-chloroacetamidine monohydrochloride, 2-chloroacetamidinehydrochloride, ethanimidamide, 2-chloro-, monohydrochloride, chloracetamidine hydrochloride, chloroacetamidinehcl
PubChem CID 2776957
IUPAC Name 2-chloroethanimidamide;hydrochloride
SMILES [H+].[Cl-].NC(=N)CCl

Specifications

Melting Point 97°C to 103°C
Quantity 25 g
UN Number UN3261
Beilstein 3678354
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 129
Percent Purity 96%
Chemical Name or Material Chloroacetamidine hydrochloride
Hazard Category H314-H317-H335
Hazard Statement GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 233-675-7
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only