missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetaldehyde Diethyl Acetal 98.0+%, TCI America™

Catalog No. C008425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C008425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C008425G Supplier TCI America Supplier No. C008425G

Chemical Identifiers

CAS 621-62-5
Molecular Formula C6H13ClO2
Molecular Weight (g/mol) 152.62
MDL Number MFCD00000949
InChI Key OVXJWSYBABKZMD-UHFFFAOYSA-N
Synonym chloroacetaldehyde diethyl acetal, chloroacetal, ethane, 2-chloro-1,1-diethoxy, diethoxyethyl chloride, 1,1-diethoxy-2-chloroethane, 2-chloroacetaldehyde diethyl acetal, monochloroacetaldehyde diethyl acetal, chloroacetaldehydediethylacetal, acetaldehyde, chloro-, diethyl acetal, 2-chloroacetal
PubChem CID 12128
IUPAC Name 2-chloro-1,1-diethoxyethane
SMILES CCOC(CCl)OCC

Specifications

Color Yellow
Boiling Point 157°C
Quantity 25 g
UN Number 1993
Formula Weight 152.62
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloroacetaldehyde Diethyl Acetal