missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetaldehyde diethyl acetal, 98%

Catalog No. AAL0766122
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0766122 100 g
AAL0766136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0766122 Supplier Thermo Scientific Chemicals Supplier No. L0766122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 621-62-5 | C6H13ClO2 | 152.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 621-62-5
Molecular Formula C6H13ClO2
Molecular Weight (g/mol) 152.62
MDL Number MFCD00000949
InChI Key OVXJWSYBABKZMD-UHFFFAOYSA-N
Synonym chloroacetaldehyde diethyl acetal, chloroacetal, ethane, 2-chloro-1,1-diethoxy, diethoxyethyl chloride, 1,1-diethoxy-2-chloroethane, 2-chloroacetaldehyde diethyl acetal, monochloroacetaldehyde diethyl acetal, chloroacetaldehydediethylacetal, acetaldehyde, chloro-, diethyl acetal, 2-chloroacetal
PubChem CID 12128
IUPAC Name 2-chloro-1,1-diethoxyethane
SMILES CCOC(CCl)OCC

Specifications

Melting Point -45°C
Density 1.018
Boiling Point 156°C to 157°C
Flash Point 50°C (122°F)
Refractive Index 1.416
Linear Formula ClCH2CH(OCH2CH3)2
Quantity 100 g
UN Number UN1993
Beilstein 969254
Sensitivity Moisture sensitive
Formula Weight 152.62
Percent Purity 98%
Chemical Name or Material Chloroacetaldehyde diethyl acetal
Show More Show Less
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 210-695-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only