missing translation for 'onlineSavingsMsg'
Learn More

Chloro-N,N,N',N'-tetramethylformamidinium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. p-7125211
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19571G 1 g
C19575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C19571G Supplier TCI America Supplier No. C19571G
Only null left

Chemical Identifiers

CAS 94790-35-9
Molecular Formula C5H12ClF6N2P
Molecular Weight (g/mol) 280.579
InChI Key CUKNPSDEURGZCO-UHFFFAOYSA-N
Synonym TCFH
PubChem CID 10989639
IUPAC Name [chloro(dimethylamino)methylidene]-dimethylazanium;hexafluorophosphate
SMILES CN(C)C(=[N+](C)C)Cl.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 107°C
Color White-Yellow
Quantity 1 g
Formula Weight 280.58
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Chloro-N,N,N′,N′-tetramethylformamidinium Hexafluorophosphate
TSCA No
Recommended Storage Freezer