missing translation for 'onlineSavingsMsg'
Learn More

α-Chloro-m-xylene, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 110160250

 View more versions of this product

Catalog No. AC110160250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

α-Chloro-m-xylene
620-19-9
100.0
195.0°C to 196.0°C
Authentic
Glass bottle
1.5335 to 1.5365
MFCD00000909
05, 373
15, 10289
Solubility in water: insoluble. Other solubilities: miscible with absolute alcohol and ether
CC1=CC(=CC=C1)CCl
140.61
140.61
96%
97.5
1.0640g/mL
75°C
97.5% min. (GC)
C8H9Cl
CH3C6H4CH2Cl
25mL
1.064
3-methylbenzyl chloride, 1-chloromethyl-3-methylbenzene, m-xylyl chloride, 3-methylbenzylchloride, alpha-chloro-m-xylene, m-methylbenzyl chloride, 1-chloromethyl-3-methyl-benzene, m-chloromethyl toluene, 3-chloromethyl toluene, benzene, 1-chloromethyl-3-methyl
LZBOHNCMCCSTJX-UHFFFAOYSA-N
1-(chloromethyl)-3-methylbenzene
12102
98%

Chemical Identifiers

620-19-9
140.61
LZBOHNCMCCSTJX-UHFFFAOYSA-N
12102
CC1=CC(=CC=C1)CCl
C8H9Cl
MFCD00000909
3-methylbenzyl chloride, 1-chloromethyl-3-methylbenzene, m-xylyl chloride, 3-methylbenzylchloride, alpha-chloro-m-xylene, m-methylbenzyl chloride, 1-chloromethyl-3-methyl-benzene, m-chloromethyl toluene, 3-chloromethyl toluene, benzene, 1-chloromethyl-3-methyl
1-(chloromethyl)-3-methylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously

GHS Signal Word: Danger

EINECSNumber : 210-628-9

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only