missing translation for 'onlineSavingsMsg'
Learn More

Chloro(dimethyl)thexylsilane 95.0+%, TCI America™

Catalog No. p-7137094
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T21165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T21165G Supplier TCI America Supplier No. T21165G

Chemical Identifiers

CAS 67373-56-2
Molecular Formula C8H19ClSi
Molecular Weight (g/mol) 178.78
MDL Number MFCD00012175
InChI Key KIGALSBMRYYLFJ-UHFFFAOYSA-N
Synonym dimethylthexylsilyl chloride, thexyldimethylchlorosilane, chloro dimethyl thexylsilane, chloro 2,3-dimethylbutan-2-yl dimethylsilane, dimethylthexylchlorosilane, thexyldimethylsilyl chloride, chlorodimethyl 1,1,2-trimethylpropyl silane, dimethylthexylsilylchloride, tds-cl, acmc-209nyu
PubChem CID 144239
IUPAC Name chloro(2,3-dimethylbutan-2-yl)dimethylsilane
SMILES CC(C)C(C)(C)[Si](C)(C)Cl

Specifications

Color Colorless
Boiling Point 56°C
Quantity 5 g
UN Number 2986
Formula Weight 178.78
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloro(dimethyl)thexylsilane