missing translation for 'onlineSavingsMsg'
Learn More

Chloro(crotyl)(tri-tert-butylphosphine)palladium(II)

Catalog No. AA4696203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4696203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4696203 Supplier Thermo Scientific Chemicals Supplier No. 04696203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1334497-00-5

Chloro(crotyl)(tri-tert-butylphosphine)palladium(II) is also used in C-C Coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1334497-00-5
MDL Number MFCD25372547
Synonym PdCl(crotyl)(PtBu3)

Specifications

Color Yellow
Quantity 1 g
Solubility Information It is insoluble in water.
Formula Weight 400.3
Physical Form Powder
Chemical Name or Material Chloro(crotyl)(tri-tert-butylphosphine)palladium(II)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only