missing translation for 'onlineSavingsMsg'
Learn More

Chloro(3-chloropropyl)dimethylsilane 96.0+%, TCI America™

Catalog No. C34265ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34265ML 5 mL
C34261ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C34265ML Supplier TCI America Supplier No. C34265ML

Chemical Identifiers

CAS 10605-40-0
Molecular Formula C5H12Cl2Si
Molecular Weight (g/mol) 171.136
MDL Number MFCD00042612
InChI Key BJLJNLUARMMMLW-UHFFFAOYSA-N
PubChem CID 82765
IUPAC Name chloro-(3-chloropropyl)-dimethylsilane
SMILES C[Si](C)(CCCCl)Cl

Specifications

Color Colorless
Boiling Point 179°C
Quantity 5 mL
UN Number 2987
Formula Weight 171.14
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloro(3-chloropropyl)dimethylsilane
EINECSNumber (2)-2047
TSCA Yes