missing translation for 'onlineSavingsMsg'
Learn More

Chloro(1,5-hexadiene)rhodium(I) Dimer 93.0+%, TCI America™

Catalog No. C3194100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3194100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C3194100MG Supplier TCI America Supplier No. C3194100MG

Chemical Identifiers

CAS 32965-49-4
Molecular Formula C12H20Cl2Rh2
MDL Number MFCD00064595
Synonym Di-mu-chlorobis[(1,2,5,6-eta)-1,5-hexadiene]dirhodium(I)

Specifications

Melting Point 120°C
Quantity 100 mg
Formula Weight 441.00
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chloro(1,5-hexadiene)rhodium(I) Dimer