missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloro(1,3-dimesitylimidazol-2-ylidene)copper(I) 97.0+%, TCI America™
$124.89 - $313.16
Chemical Identifiers
| CAS | 873779-78-3 |
|---|---|
| Molecular Formula | C21H24ClCuN2 |
| Synonym | [1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene]chlorocopper(I) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C2422200MG
|
TCI America
C2422200MG |
200 mg |
Each for $124.89
|
|
|||||
|
C24221G
|
TCI America
C24221G |
1 g |
Each for $313.16
|
|
|||||
Chemical Identifiers
| 873779-78-3 | |
| C21H24ClCuN2 | |
| [1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene]chlorocopper(I) |
Specifications
| 873779-78-3 | |
| C21H24ClCuN2 | |
| [1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene]chlorocopper(I) | |
| ≥97.0% (T) | |
| Chloro(1,3-dimesitylimidazol-2-ylidene)copper(I) |
| 277°C | |
| 200 mg | |
| 403.43 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer