missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlormethiazole Hydrochloride 98.0+%, TCI America™
$357.90 - $859.33
Chemical Identifiers
| CAS | 6001-74-7 |
|---|---|
| Molecular Formula | C6H9Cl2NS |
| Molecular Weight (g/mol) | 198.11 |
| MDL Number | MFCD00673948 |
| InChI Key | OFXYKSLKNMTBHK-UHFFFAOYSA-N |
| Synonym | chlormethiazole hydrochloride, clomethiazole hydrochloride, somnevrin hydrochloride, 5-2-chloroethyl-4-methylthiazole hydrochloride, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride, 2-4-methylthiazol-5-yl ethyl chloride hydrochloride, clomethiazole hydrochloride mi, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride 1:1, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride 1:1 |
| PubChem CID | 19035073 |
| IUPAC Name | 5-(2-chloroethyl)-4-methyl-1,3-thiazole hydrochloride |
| SMILES | Cl.CC1=C(CCCl)SC=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C3020200MG
|
TCI America
C3020200MG |
200 mg |
Each for $357.90
|
|
|||||
|
C30201G
|
TCI America
C30201G |
1 g |
Each for $859.33
|
|
|||||
Chemical Identifiers
| 6001-74-7 | |
| 198.11 | |
| OFXYKSLKNMTBHK-UHFFFAOYSA-N | |
| 19035073 | |
| Cl.CC1=C(CCCl)SC=N1 |
| C6H9Cl2NS | |
| MFCD00673948 | |
| chlormethiazole hydrochloride, clomethiazole hydrochloride, somnevrin hydrochloride, 5-2-chloroethyl-4-methylthiazole hydrochloride, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride, 2-4-methylthiazol-5-yl ethyl chloride hydrochloride, clomethiazole hydrochloride mi, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride 1:1, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride 1:1 | |
| 5-(2-chloroethyl)-4-methyl-1,3-thiazole hydrochloride |
Specifications
| 6001-74-7 | |
| White | |
| MFCD00673948 | |
| chlormethiazole hydrochloride, clomethiazole hydrochloride, somnevrin hydrochloride, 5-2-chloroethyl-4-methylthiazole hydrochloride, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride, 2-4-methylthiazol-5-yl ethyl chloride hydrochloride, clomethiazole hydrochloride mi, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride 1:1, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride 1:1 | |
| Cl.CC1=C(CCCl)SC=N1 | |
| 198.11 | |
| 198.11 | |
| Crystalline Powder |
| 136°C | |
| C6H9Cl2NS | |
| 200 mg | |
| OFXYKSLKNMTBHK-UHFFFAOYSA-N | |
| 5-(2-chloroethyl)-4-methyl-1,3-thiazole hydrochloride | |
| 19035073 | |
| ≥98.0% (HPLC,T) | |
| Chlormethiazole Hydrochloride |
Safety and Handling
TSCA : No