missing translation for 'onlineSavingsMsg'
Learn More

Chlormadinone Acetate 97.0+%, TCI America™

Catalog No. c31161g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C31161G Supplier TCI America Supplier No. C31161G
Only null left

Specifications

CAS 302-22-7
Melting Point 208°C
Color White-Yellow
Molecular Formula C23H29ClO4
Quantity 1 g
MDL Number MFCD00056471
Synonym 17alpha-Acetoxy-6-chloro-6-dehydroprogesterone, 6-Chloro-17alpha-hydroxypregna-4,6-diene-3,20-dione Acetate, 17alpha-Acetoxy-6-chloropregna-4,6-diene-3,20-dione
InChI Key QMBJSIBWORFWQT-DFXBJWIESA-N
SMILES CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(Cl)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)=O
IUPAC Name (1R,3aS,3bR,9aR,9bS,11aS)-1-acetyl-5-chloro-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl acetate
Molecular Weight (g/mol) 404.93
PubChem CID 9324
ChEBI CHEBI:31394
Formula Weight 404.93
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Chlormadinone Acetate
Show More Show Less
EINECSNumber (9)-2348
RTECSNumber TU3750000
TSCA No
Recommended Storage Freezer