missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Chlorine High Performance Pipe Marker, 0.375 to 0.5 in.

Catalog No. 18038228
Change view
Click to view available options
Color:
Black on Orange
Black on Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18038228 Black on Yellow
18038229 Black on Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18038228 Supplier Brady™ Supplier No. CHLORINEOO

Wrap Around High Performance Pipe Markers (B-689) are coiled, printed plastic sheets with sealed color graphics for use on pipes in harsh environments.

  • Designed for use on rusty, dirty, wet or rough pipes where pressure sensitive labels aren’t effective.
  • Resistant to UV light exposure, high humidity, high temperatures, abrasion, chemical reagents and physical agitation.
  • Self-adhesive end strip adheres to the marker's body to ensure a tight, continuous wrap.
  • Excellent chemical resistance.

Specifications

Adhesive Type Non-adhesive
Coating Laminate
Color Black on Yellow
Display Text
Finish Gloss
Format Sheet
Material Coiled Plastic (B-689)
Product Type Pipe Marker
Sterility Non-sterile
Dimensions (L x W) 4 x 4 in.
Length (English) 4 in.
Width (English) 4 in.
Width (Metric) 101.60 mm
Show More Show Less