missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LaMotte™ Chlorine DPD Tablet No. 1
Description
- 100 tablets per box.
Specifications
Specifications
| Color | White |
| Composition | 6283-63-2 |
| Description | N,N diethyl-p-phenylenediamine sulfate |
| Format | Solid |
| For Use With (Equipment) | LaMotte Colorimeters |
| Packaging | Box |
| Quantity | 100 Pk. |
| Type | Chlorine DPD Tablet No. 1 |