missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorhexidine Dihydrochloride 96.0+%, TCI America™
$108.11 - $260.97
Chemical Identifiers
| CAS | 3697-42-5 |
|---|---|
| Molecular Formula | C22H32Cl4N10 |
| Molecular Weight (g/mol) | 578.368 |
| MDL Number | MFCD00068998 |
| InChI Key | WJLVQTJZDCGNJN-UHFFFAOYSA-N |
| Synonym | 1,1′C-Hexamethylenebis[5-(4-chlorophenyl)biguanide] Dihydrochloride |
| PubChem CID | 9571016 |
| IUPAC Name | (1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine;dihydrochloride |
| SMILES | C1=CC(=CC=C1NC(=NC(=NCCCCCCN=C(N)N=C(N)NC2=CC=C(C=C2)Cl)N)N)Cl.Cl.Cl |
Chemical Identifiers
| 3697-42-5 | |
| 578.368 | |
| WJLVQTJZDCGNJN-UHFFFAOYSA-N | |
| 9571016 | |
| C1=CC(=CC=C1NC(=NC(=NCCCCCCN=C(N)N=C(N)NC2=CC=C(C=C2)Cl)N)N)Cl.Cl.Cl |
| C22H32Cl4N10 | |
| MFCD00068998 | |
| 1,1′C-Hexamethylenebis[5-(4-chlorophenyl)biguanide] Dihydrochloride | |
| (1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine;dihydrochloride |
Specifications
| 3697-42-5 | |
| White-Yellow | |
| MFCD00068998 | |
| 3077 | |
| WJLVQTJZDCGNJN-UHFFFAOYSA-N | |
| (1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine;dihydrochloride | |
| 9571016 | |
| ≥96.0% (T) | |
| Chlorhexidine Dihydrochloride |
| 134°C | |
| C22H32Cl4N10 | |
| 5 g | |
| 1,1′C-Hexamethylenebis[5-(4-chlorophenyl)biguanide] Dihydrochloride | |
| C1=CC(=CC=C1NC(=NC(=NCCCCCCN=C(N)N=C(N)NC2=CC=C(C=C2)Cl)N)N)Cl.Cl.Cl | |
| 578.368 | |
| 578.37 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : XA5285000
TSCA : No