Learn More
Chlorhexidine digluconate, 20% w/v aq. soln., non-sterile
CAS: 18472-51-0 | C34H54Cl2N10O14 | 897.76 g/mol
Supplier: Thermo Scientific Chemicals 041385AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
ApplicationsChlorhexidine digluconate, 20% w/v aq. Soln is used as an oral antiseptic agent and anti-microbial agent. It is an active ingredient in germicidal mouthwash, pet shampoo, surgical lubricants and wipes and hand soap. It is utilized in the study to know the role of essential oils for improving skin antisepsis. It plays a vital role as disinfectants and preservative in pharmaceutical products. It is also used as an additive in cosmetics such as creams, toothpaste and deodorants.
Solubility
Miscible with water, ethanol and acetone.
Notes
Store in a cool place. Light sensitive. Incompatible with strong oxidizing agents.
Specifications
| Chlorhexidine digluconate | |
| 18472-51-0 | |
| 1.06 g/mL | |
| C34H54Cl2N10O14 | |
| 100 mL | |
| peridex, periogard, chlorhexidine gluconate, 1,1'-hexamethylene bis 5-p-chlorophenyl biguanide di-d-gluconate | |
| OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1 | |
| 897.76 | |
| 897.76 | |
| Liquid |
| Non-sterile | |
| Colorless | |
| 20% w/v aq. soln. | |
| MFCD00083599 | |
| Light sensitive | |
| YZIYKJHYYHPJIB-UUPCJSQJSA-N | |
| (1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid;sulfane | |
| 60148207 | |
| 20% w/v aq. soln. |
Chemical Identifiers
| 18472-51-0 | |
| 897.76 | |
| YZIYKJHYYHPJIB-UUPCJSQJSA-N | |
| 60148207 | |
| OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1 |
| C34H54Cl2N10O14 | |
| MFCD00083599 | |
| peridex, periogard, chlorhexidine gluconate, 1,1'-hexamethylene bis 5-p-chlorophenyl biguanide di-d-gluconate | |
| (1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid;sulfane |
Safety and Handling
P280-P305+P351+P338-P310
H318
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 242-354-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only