missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorhexidine Diacetate 98.0+%, TCI America™
$84.70 - $256.05
Chemical Identifiers
| CAS | 56-95-1 |
|---|---|
| Molecular Formula | C26H38Cl2N10O4 |
| Molecular Weight (g/mol) | 625.56 |
| MDL Number | MFCD00150042 |
| InChI Key | WDRFFJWBUDTUCA-UHFFFAOYSA-N |
| Synonym | chlorhexidine acetate hydrate 1:2:x, chlorhexidine diacetate salt monohydrate, chlorhexidine diacetate monohydrate, 1,1 inverted exclamation marka-hexamethylenebis 5-4-chlorophenyl biguanide diacetate salt, 4-chlorophenyl 6-4-chlorophenyl amino iminomethyl amino iminomethy l amino hexyl amino iminomethyl amino iminomethyl amine, acetic acid, acetic a cid, oxamethane |
| PubChem CID | 43834499 |
| IUPAC Name | N'-(6-{[amino({amino[(4-chlorophenyl)amino]methylidene}amino)methylidene]amino}hexyl)-1-({amino[(4-chlorophenyl)amino]methylidene}amino)methanimidamide; bis(acetic acid) |
| SMILES | CC(O)=O.CC(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1 |
Chemical Identifiers
| 56-95-1 | |
| 625.56 | |
| WDRFFJWBUDTUCA-UHFFFAOYSA-N | |
| 43834499 | |
| CC(O)=O.CC(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1 |
| C26H38Cl2N10O4 | |
| MFCD00150042 | |
| chlorhexidine acetate hydrate 1:2:x, chlorhexidine diacetate salt monohydrate, chlorhexidine diacetate monohydrate, 1,1 inverted exclamation marka-hexamethylenebis 5-4-chlorophenyl biguanide diacetate salt, 4-chlorophenyl 6-4-chlorophenyl amino iminomethyl amino iminomethy l amino hexyl amino iminomethyl amino iminomethyl amine, acetic acid, acetic a cid, oxamethane | |
| N'-(6-{[amino({amino[(4-chlorophenyl)amino]methylidene}amino)methylidene]amino}hexyl)-1-({amino[(4-chlorophenyl)amino]methylidene}amino)methanimidamide; bis(acetic acid) |
Specifications
| White | |
| 56-95-1 | |
| C26H38Cl2N10O4 | |
| 3077 | |
| 5 g | |
| CC(O)=O.CC(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1 | |
| 625.56 | |
| 625.56 | |
| Chlorhexidine Diacetate |
| Crystal-Powder at 20°C | |
| 155°C | |
| MFCD00150042 | |
| chlorhexidine acetate hydrate 1:2:x, chlorhexidine diacetate salt monohydrate, chlorhexidine diacetate monohydrate, 1,1 inverted exclamation marka-hexamethylenebis 5-4-chlorophenyl biguanide diacetate salt, 4-chlorophenyl 6-4-chlorophenyl amino iminomethyl amino iminomethy l amino hexyl amino iminomethyl amino iminomethyl amine, acetic acid, acetic a cid, oxamethane | |
| WDRFFJWBUDTUCA-UHFFFAOYSA-N | |
| N'-(6-{[amino({amino[(4-chlorophenyl)amino]methylidene}amino)methylidene]amino}hexyl)-1-({amino[(4-chlorophenyl)amino]methylidene}amino)methanimidamide; bis(acetic acid) | |
| 43834499 | |
| ≥98.0% (T) |
Safety and Handling
RTECSNumber : DU1930000
TSCA : No