missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine Diacetate 98.0+%, TCI America™

Catalog No. C251125G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C251125G 25 g
C25115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C251125G Supplier TCI America Supplier No. C251125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56-95-1
Molecular Formula C26H38Cl2N10O4
Molecular Weight (g/mol) 625.56
MDL Number MFCD00150042
InChI Key WDRFFJWBUDTUCA-UHFFFAOYSA-N
Synonym chlorhexidine acetate hydrate 1:2:x, chlorhexidine diacetate salt monohydrate, chlorhexidine diacetate monohydrate, 1,1 inverted exclamation marka-hexamethylenebis 5-4-chlorophenyl biguanide diacetate salt, 4-chlorophenyl 6-4-chlorophenyl amino iminomethyl amino iminomethy l amino hexyl amino iminomethyl amino iminomethyl amine, acetic acid, acetic a cid, oxamethane
PubChem CID 43834499
IUPAC Name N'-(6-{[amino({amino[(4-chlorophenyl)amino]methylidene}amino)methylidene]amino}hexyl)-1-({amino[(4-chlorophenyl)amino]methylidene}amino)methanimidamide; bis(acetic acid)
SMILES CC(O)=O.CC(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1

Specifications

Color White
Physical Form Crystal-Powder at 20°C
Melting Point 155°C
UN Number 3077
Quantity 25 g
Formula Weight 625.56
Percent Purity ≥98.0% (T)
Chemical Name or Material Chlorhexidine Diacetate
RTECSNumber DU1930000
TSCA No