missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine diacetate hydrate, 98%

Catalog No. AC214980050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC214980050 5 g Glass bottle
AC214980250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC214980050 Supplier Thermo Scientific Chemicals Supplier No. 214980050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 206986-79-0 | C26H38Cl2N10O4 | 625.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 206986-79-0
Molecular Formula C26H38Cl2N10O4
Molecular Weight (g/mol) 625.56
MDL Number MFCD00150042
InChI Key WDRFFJWBUDTUCA-UHFFFAOYSA-N
Synonym chlorhexidine acetate hydrate 1:2:x, chlorhexidine diacetate salt monohydrate, chlorhexidine diacetate monohydrate, 1,1 inverted exclamation marka-hexamethylenebis 5-4-chlorophenyl biguanide diacetate salt, 4-chlorophenyl 6-4-chlorophenyl amino iminomethyl amino iminomethy l amino hexyl amino iminomethyl amino iminomethyl amine, acetic acid, acetic a cid, oxamethane
PubChem CID 43834499
SMILES CC(O)=O.CC(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1

Specifications

Color White
Physical Form Powder
Melting Point 153.0°C to 157.0°C
Assay Percent Range 98%
Solubility Information Solubility in water: 19g/L (20°C). Other solubilities: soluble in alcohol, glycerol, propylene glycol and, polyethylene glycols
Quantity 5 g
IUPAC Name acetic acid;(1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine;hydrate
Formula Weight 625.55
Percent Purity 98%
Infrared Spectrum Authentic
Loss on Drying 3.5% max. (105°C)
Packaging Glass bottle
Chemical Name or Material Chlorhexidine diacetate hydrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 200-302-4
RTECSNumber DU1930000

RUO – Research Use Only