missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorendic anhydride, 95%, may cont. up to 3% chlorendic acid
Description
Chlorendic anhydride is used in the synthesis of polymers, used in production of fiberglass-reinforced resins for chemical industry equipment. It can be used to make alkyd resins for use in special inks and paints. It is used as a hardening agent in epoxy resins used in manufacture of printed circuit boards
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorendic anhydride is used in the synthesis of polymers, used in production of fiberglass-reinforced resins for chemical industry equipment. It can be used to make alkyd resins for use in special inks and paints. It is used as a hardening agent in epoxy resins used in manufacture of printed circuit boards
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chlorendic anhydride is used in the synthesis of polymers, used in production of fiberglass-reinforced resins for chemical industry equipment. It can be used to make alkyd resins for use in special inks and paints. It is used as a hardening agent in epoxy resins used in manufacture of printed circuit boards
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 235°C to 239°C |
| Density | 1.73 |
| Quantity | 50 g |
| Beilstein | 92693 |
| Merck Index | 14,2086 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 370.83 |
| Percent Purity | 96% |
| Assay | may cont. up to 3% chlorendic acid |
| Chemical Name or Material | Chlorendic anhydride |
Safety and Handling
| Hazard Category | H315-H319-H335-H350 |
| Hazard Statement | GHS H Statement H350-H315-H319-H335 May cause cancer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c |
| EINECSNumber | 204-077-3 |
| RTECSNumber | RB9080000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only