missing translation for 'onlineSavingsMsg'
Learn More

Chlorazol Black E, TCI America™

Catalog No. C053325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C053325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C053325G Supplier TCI America Supplier No. C053325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory reagent grade dye suitable for research and analytical applications.

Chemical Identifiers

CAS 1937-37-7
Molecular Formula C34H25N9Na2O7S2
Molecular Weight (g/mol) 781.73
MDL Number MFCD00066363
InChI Key OLVNAWLXZDRGPL-UHFFFAOYSA-L
Synonym Direct Black 38
PubChem CID 44135447
IUPAC Name disodium;4-amino-3-[[4-[4-[(2,4-diaminophenyl)diazenyl]phenyl]phenyl]diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)C5=CC=C(C=C5)N=NC6=C(C=C(C=C6)N)N)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Black
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 781.73
Grade Reagent
Chemical Name or Material Chlorazol Black E
EINECSNumber (5)-1370
RTECSNumber QJ6160000
TSCA Yes