missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Chlorazol Black E

Supplier:  Thermo Scientific™ A1325909

 View more versions of this product

Catalog No. AAA1325909


Only null left

Description

Description

Specifications

Specifications

Chlorazol Black E
C34H25N9Na2O7S2
Azo Black; C.I. 30235
C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)C5=CC=C(C=C5)N=NC6=C(C=C(C=C6)N)N)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
781.73
781.73
1937-37-7
MFCD00066363
OLVNAWLXZDRGPL-UHFFFAOYSA-L
disodium;4-amino-3-[[4-[4-[(2,4-diaminophenyl)diazenyl]phenyl]phenyl]diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
44135447
10 g

Chemical Identifiers

1937-37-7
781.73
OLVNAWLXZDRGPL-UHFFFAOYSA-L
44135447
C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)C5=CC=C(C=C5)N=NC6=C(C=C(C=C6)N)N)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
C34H25N9Na2O7S2
MFCD00066363
Azo Black; C.I. 30235
disodium;4-amino-3-[[4-[4-[(2,4-diaminophenyl)diazenyl]phenyl]phenyl]diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H361
May cause cancer.
Suspected of damaging fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H350-H361d

EINECSNumber : 217-710-3

RTECSNumber : QJ6160000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only