missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Chlorazol Black E

Catalog No. AAA1325918
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1325918 50 g
AAA1325909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1325918 Supplier Thermo Scientific™ Supplier No. A1325918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1937-37-7
Molecular Formula C34H25N9Na2O7S2
Molecular Weight (g/mol) 781.73
MDL Number MFCD00066363
InChI Key OLVNAWLXZDRGPL-UHFFFAOYSA-L
Synonym Azo Black; C.I. 30235
PubChem CID 44135447
IUPAC Name disodium;4-amino-3-[[4-[4-[(2,4-diaminophenyl)diazenyl]phenyl]phenyl]diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)C5=CC=C(C=C5)N=NC6=C(C=C(C=C6)N)N)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Quantity 50 g
Formula Weight 781.73
Chemical Name or Material Chlorazol Black E
Hazard Category H350-H361d
Hazard Statement GHS H Statement
H350-H361
May cause cancer.
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 217-710-3
RTECSNumber QJ6160000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only