missing translation for 'onlineSavingsMsg'
Learn More

Chloranilic Acid Lanthanum(III) Salt Decahydrate 98.0+%, TCI America™

Catalog No. C00795G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C00795G 5 g
C007925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C00795G Supplier TCI America Supplier No. C00795G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity crystalline powder for laboratory and research use.

  • Crystalline powder prepared from chelometric titration
  • Confirmed structural analysis by IR stored under room temperature

Chemical Identifiers

CAS 32607-23-1
Molecular Formula C18H6Cl6La2O12
Molecular Weight (g/mol) 904.745
MDL Number MFCD00135764
InChI Key GGKWUCDKAJOTCW-UHFFFAOYSA-N
Synonym chloranilic acid lanthanum salt, chloranilic acid, lanthanum salt
PubChem CID 126959841
IUPAC Name 2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione;lanthanum
SMILES C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O.C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O.C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O.[La].[La]

Specifications

Color Black
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range >98.0%
Formula Weight 898.70
Percent Purity ≥98.0% (T)
Appearance Black Powder to Crystal
Chemical Name or Material Chloranilic Acid Lanthanum(III) Salt Decahydrate