Learn More
Chloranilic acid, 98+%
CAS: 87-88-7 | C6H2Cl2O4 | 208.978 g/mol
Supplier: Thermo Scientific Chemicals A1049322
Description
Chloranilic acid is used in spectrophotometry. It reacts with metal ions to form stable complexes. It is also used as a reactant in the preparation of dimethylbipyridyl complexes and (nonylbenzimidazolylmethyl)benzene. Further, it is used in charge-transfer reactions with metoprolol tartrate. It is also employed in the synthesis of osmium metalla-rectangles with anticancer activity. In addition to this, it has potential antibacterial activities against methicillin-resistant Staphylococcus aureus.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChloranilic acid is used in spectrophotometry. It reacts with metal ions to form stable complexes. It is also used as a reactant in the preparation of dimethylbipyridyl complexes and (nonylbenzimidazolylmethyl)benzene. Further, it is used in charge-transfer reactions with metoprolol tartrate. It is also employed in the synthesis of osmium metalla-rectangles with anticancer activity. In addition to this, it has potential antibacterial activities against methicillin-resistant Staphylococcus aureus.
Solubility
Soluble in hot water and methanol. Insoluble in organic solvents.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Chloranilic acid | |
| >300°C (decomposition) | |
| MFCD00001596 | |
| 1875040 | |
| chloranilic acid, 2,5-dichloro-3,6-dihydroxy-p-benzoquinone, p-chloranilic acid, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro-3,6-dihydroxy, unii-yj8l3bb7y4, 2,5-dichloro-3,6-dihydroxybenzoquinone, 2,5-dihydroxy-3,6-dichlorobenzoquinone, yj8l3bb7y4, p-quinone, 2,5-dichloro-3,6-dihydroxy, p-benzoquinone, 2,5-dichloro-3,6-dihydroxy | |
| IPPWILKGXFOXHO-UHFFFAOYSA-N | |
| 2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione | |
| 66604 | |
| ≥98% |
| 87-88-7 | |
| C6H2Cl2O4 | |
| 100 g | |
| 14,2079 | |
| Soluble in hot water and methanol. Insoluble in organic solvents. | |
| C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O | |
| 208.978 | |
| 208.99 |
Chemical Identifiers
| 87-88-7 | |
| 208.978 | |
| IPPWILKGXFOXHO-UHFFFAOYSA-N | |
| 66604 | |
| C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O |
| C6H2Cl2O4 | |
| MFCD00001596 | |
| chloranilic acid, 2,5-dichloro-3,6-dihydroxy-p-benzoquinone, p-chloranilic acid, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro-3,6-dihydroxy, unii-yj8l3bb7y4, 2,5-dichloro-3,6-dihydroxybenzoquinone, 2,5-dihydroxy-3,6-dichlorobenzoquinone, yj8l3bb7y4, p-quinone, 2,5-dichloro-3,6-dihydroxy, p-benzoquinone, 2,5-dichloro-3,6-dihydroxy | |
| 2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 201-780-7
RTECSNumber : DK4005000
TSCA : Yes
RUO – Research Use Only