missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloranil 98.0+%, TCI America™
$44.73 - $218.10
Chemical Identifiers
| CAS | 118-75-2 |
|---|---|
| Molecular Formula | C6Cl4O2 |
| Molecular Weight (g/mol) | 245.86 |
| MDL Number | MFCD00001594 |
| InChI Key | UGNWTBMOAKPKBL-UHFFFAOYSA-N |
| Synonym | chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone |
| PubChem CID | 8371 |
| ChEBI | CHEBI:36703 |
| IUPAC Name | tetrachlorocyclohexa-2,5-diene-1,4-dione |
| SMILES | ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O |
Chemical Identifiers
| 118-75-2 | |
| 245.86 | |
| UGNWTBMOAKPKBL-UHFFFAOYSA-N | |
| 8371 | |
| tetrachlorocyclohexa-2,5-diene-1,4-dione |
| C6Cl4O2 | |
| MFCD00001594 | |
| chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone | |
| CHEBI:36703 | |
| ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O |
Specifications
| 118-75-2 | |
| Yellow | |
| MFCD00001594 | |
| 3077 | |
| UGNWTBMOAKPKBL-UHFFFAOYSA-N | |
| tetrachlorocyclohexa-2,5-diene-1,4-dione | |
| 8371 | |
| 245.86 | |
| Crystalline Powder |
| 297°C | |
| C6Cl4O2 | |
| 25 g | |
| chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone | |
| ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O | |
| 245.86 | |
| CHEBI:36703 | |
| ≥98.0% (GC,T) | |
| Chloranil |
Safety and Handling
EINECSNumber : (3)-1007
RTECSNumber : DK6825000
TSCA : Yes