missing translation for 'onlineSavingsMsg'
Learn More

Chloranil (ca. 2% in N,N-Dimethylformamide) [for Detection of Primary and Secondary Amines], TCI America™

Catalog No. C177010ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C177010ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C177010ML Supplier TCI America Supplier No. C177010ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 118-75-2
Molecular Formula C6Cl4O2
Molecular Weight (g/mol) 245.86
MDL Number MFCD00001594
InChI Key UGNWTBMOAKPKBL-UHFFFAOYSA-N
Synonym chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone
PubChem CID 8371
ChEBI CHEBI:36703
IUPAC Name tetrachlorocyclohexa-2,5-diene-1,4-dione
SMILES ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O

Specifications

Quantity 10 mL
UN Number 2265
Formula Weight 245.86
Physical Form Liquid
Chemical Name or Material Chloranil (ca. 2% in N,N-Dimethylformamide) [for Detection of Primary and Secondary Amines]
EINECSNumber (3)-1007
RTECSNumber DK6825000
TSCA Yes