missing translation for 'onlineSavingsMsg'
Learn More

Chitosan, Powder, Spectrum™ Chemical

Catalog No. p-7157183
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601571 25 g Poly Bottle
18601573 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18601571 Supplier Spectrum Chemical Mfg Cor Supplier No. C156925GM
Only null left

C1569, 9012-76-4, (C6H11NO4)n

Spectrum™ Chemical Chitosan, Powder is a polysaccharide made by treating shrimp and other crustacean shells with the alkali sodium hydroxide. It can be used in agriculture as a seed treatment and to fight off fungal infections and is often used in the wine making industry as a chemical to prevent spoilage. In medicine it can be used in bandages to reduce bleeding and as an antibacterial agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 9012-76-4
Molecular Formula C56H103N9O39
Molecular Weight (g/mol) 1526.46
InChI Key FLASNYPZGWUPSU-SICDJOISSA-N
IUPAC Name methyl N-[(2S,3R,4R,5S,6R)-5-{[(2S,3R,4R,5S,6R)-3-amino-5-{[(2S,3R,4R,5S,6R)-3-amino-5-{[(2S,3R,4R,5S,6R)-3-amino-5-{[(2S,3R,4R,5S,6R)-3-amino-5-{[(2S,3R,4R,5S,6R)-3-amino-5-{[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2-{[(2R,3S,4R,5R,6S)-5-amino-6-{[(2R,3S,4R,5R,6R)-5-amino-4,6-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-4-hydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-3-yl]carbamate
SMILES COC(=O)N[C@@H]1[C@@H](O)[C@H](O[C@@H]2O[C@H](CO)[C@@H](O[C@@H]3O[C@H](CO)[C@@H](O[C@@H]4O[C@H](CO)[C@@H](O[C@@H]5O[C@H](CO)[C@@H](O[C@@H]6O[C@H](CO)[C@@H](O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7N)[C@H](O)[C@H]6N)[C@H](O)[C@H]5N)[C@H](O)[C@H]4N)[C@H](O)[C@H]3N)[C@H](O)[C@H]2N)[C@@H](CO)O[C@H]1O[C@H]1[C@H](O)[C@@H](N)[C@H](O[C@H]2[C@H](O)[C@@H](N)[C@H](O)O[C@@H]2CO)O[C@@H]1CO

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 25 g
Grade Reagent