missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Chitosan

Catalog No. 448877250G
Encompass
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
448877250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 448877250G Supplier Sigma Aldrich Supplier No. 448877250G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Forms gels with multivalent anions. Gives clear solutions that dry to strong, clear films.

Specifications

CAS 9012-76-4
Quantity 250 g
MDL Number MFCD00161512
Synonym Deacetylated chitin; Poly(D-glucosamine)