missing translation for 'onlineSavingsMsg'
Learn More

Chitin, TCI America™

Catalog No. C007225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C007225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C007225G Supplier TCI America Supplier No. C007225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1398-61-4
Molecular Formula (C8H13NO5)nC16H28N2O11
MDL Number MFCD00466914
Synonym chitin from shrimp shells, n-2r-2,4,5-trihydroxy-6-hydroxymethyl oxan-3-yl acetamide, chitin from shrimp shells, practical grade, powder, chitin from shrimp shells, vetec tm reagent grade, chitin from shrimp shells, practical grade, coarse flakes, chitin from shrimp shells, bioreagent, suitable for analysis of chitinase, purified powder

Specifications

Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Chitin
RTECSNumber FM6125000
TSCA Yes