missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ Chill-Its™ 8935MF Evaporative Class Headwear Hi-Vis Ranger Hat with Microfiber Lining
Ranger hat with wide brim and vented mesh side panels is lined with an advanced microfiber material for cooling relief.
$28.86 - $28.86
Specifications
| Material | Microfiber-lined Polyester |
|---|---|
| Product Type | Ranger Hat |
| Color | Lime |
| For Use With (Application) | Road Construction, Utility Crew, Construction |
| Catalog Number | Mfr. No. | Price | Quantity | ||||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Price | Quantity | ||||||
|
19-819-583
|
Ergodyne™
12595 |
Each for $28.86
|
|
||||||
Description
- Microfiber material soft and absorbent
- Feels significantly cooler than ambient air
- Easily activated by running under water for one minute
- ANSI certified 150D Oxford Polyester
- ANSI-compliant tape with contrasting color
Specifications
| Microfiber-lined Polyester | |
| Ranger Hat | |
| Lime | |
| Road Construction, Utility Crew, Construction |