missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6716 Evaporative Cooling Hard Hat Liner

Catalog No. 19819503
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819503 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819503 Supplier Ergodyne™ Supplier No. ERG12338

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill-Its™ 6716 Evaporative Cooling Hard Hat Liner attaches to standard hard hat headbands.

  • Lightweight, cooling acrylic polymers
  • Hook and loop closures
  • Conveniently attaches to hard hat suspension headband
  • Soak in water 2 to 5 minutes to activate
  • Remains hydrated for up to 4 hours
  • Reactivate by re-soaking

Specifications

Color Blue
Closure Type Hook and Loop
For Use With (Application) Construction, Maintenance, Landscaping, Warehousing/Distribution