missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ Chill-Its™ 6710CT Evaporative Cooling Triangle Hat with Cooling Towel
Description
- Advanced PVA material activates easily and quickly once run under water for one minute
- Reusable
- Super-evaporative cooling process helps keep you cool
- Lightweight and low profile
- Tie closure
- Available in a variety of patterns and colors like blue, lime, red western, blue western, stars and stripes, flames, camouflage, and skulls
Specifications
Specifications
| Product Type | Cooling Hat |
| Color | Red, White, Blue |
| Closure Type | Tie |
| For Use With (Application) | Construction, Maintenance, Landscaping, Material Handling |
| Includes | Cooling Towel |