missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ Chill-Its™ 6710 Evaporative Cooling Triangle Hat
Description
- Activate by soaking in water for 2-5 minutes
- Reusable
- Tie closure
- Available in a variety of colors and patterns like stars and stripes, camouflage, red western, navy western, blue, lime, flames, and skulls
Specifications
Specifications
| Product Type | Cooling Hat |
| Color | Red, White, Blue |
| Closure Type | Tie |
| For Use With (Application) | Construction, Maintenance, Landscaping, Material Handling |