missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6705CT Evaporative Hook and Loop Bandana with Cooling Towel

Catalog No. 19819564
Change view
Click to view available options
Color:
Black
Blue
Blue, White
Lime
Red, White
Red, White, Blue
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819564 Black
19819556 Red, White, Blue
19819558 Red, White
19819559 Blue, White
19819560 Lime
19819561 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19819564 Supplier Ergodyne™ Supplier No. 12580

Chill-Its™ 6705CT Evaporative Hook and Loop Bandana with Cooling Towel features advanced PVA material that keeps you cool without the bulk.

  • Advanced PVA material activates easily and quickly when run under water for one minute
  • Reusable
  • Super-evaporative cooling process helps keep you cool
  • Breakaway hook and loop closure
  • Available in a variety of colors and patterns like red western, blue western, camo, stars and striped, black, lime, blue, skulls, and flames

Specifications

Material PVA
Product Type Bandana
Color Black
Closure Type Hook and Loop
For Use With (Application) Construction, Maintenance, Construction, Landscaping, Material Handling
Includes Cooling Towel