missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6705 Evaporative Cooling Bandana with Hook and Loop

Catalog No. p-7163098
Change view
Click to view available options
Color:
Black
Blue, White
Camouflage
Lime
Red, White
Red, White, Blue
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819476 Red, White, Blue
19819483 Camouflage
19819484 Red, White
19819485 Blue, White
19819486 Lime
19819497 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19819476 Supplier Ergodyne™ Supplier No. 12313
Only null left

Evaporative Cooling Bandana features activated polymers that bring cooling power to a variety of application.

  • Soak in water 2-5 minutes to activate
  • Remains hydrated for up to 4 hours
  • Reusable
  • Available in a variety of colors and patterns like red western, blue western, camo, stars and striped, black, lime, blue, skulls, and flames

Specifications

Product Type Bandana
Color Red, White, Blue
Closure Type Hook and Loop
For Use With (Application) Construction, Maintenance, Landscaping, Material Handling