missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ Chill-Its™ 6700CT Evaporative Cooling Tie Bandana with Cooling Towel
Description
- Advanced PVA material activates easily and quickly when run under water for one minute
- Reusable
- Super-evaporative cooling process helps keep you cool
- Lightweight and low profile
- Available in a variety of colors and patterns like blue, hi-vis lime, red western, blue western, stars and stripes, flames, camouflage, and skulls
Specifications
Specifications
| Material | PVA |
| Product Type | Bandana |
| Color | Red, White, Blue |
| Closure Type | Tie |
| For Use With (Application) | Construction, Maintenance, Landscaping, Material Handling |