missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6700CT Evaporative Cooling Tie Bandana with Cooling Towel

Catalog No. 19819546
Change view
Click to view available options
Color:
Black
Blue
Blue, White
Camouflage
Gray, Black, White
Lime
Red, Orange, Black
Red, White
Red, White, Blue
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819546 Red, White, Blue
19819547 Camouflage
19819548 Red, White
19819550 Blue, White
19819551 Black
19819552 Lime
296366992 Blue
19819554 Red, Orange, Black
19819555 Gray, Black, White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19819546 Supplier Ergodyne™ Supplier No. 12561

Evaporative Cooling Bandana with Cooling Towel features advanced PVA material, which keeps you cool without any bulk.

  • Advanced PVA material activates easily and quickly when run under water for one minute
  • Reusable
  • Super-evaporative cooling process helps keep you cool
  • Lightweight and low profile
  • Available in a variety of colors and patterns like blue, hi-vis lime, red western, blue western, stars and stripes, flames, camouflage, and skulls

Specifications

Material PVA
Product Type Bandana
Color Red, White, Blue
Closure Type Tie
For Use With (Application) Construction, Maintenance, Landscaping, Material Handling