missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6700 Evaporative Cooling Tie Bandana

Catalog No. p-7163095
Change view
Click to view available options
Color:
Black
Camouflage
Gray, Black, White
Red, White, Blue
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819474 Red, White, Blue
19819475 Camouflage
19819494 Gray, Black, White
19819496 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19819474 Supplier Ergodyne™ Supplier No. 12303
Only null left

Evaporative Cooling Bandana features activated polymers that bring cooling power to a variety of application.

  • Activate by soaking in water for 2-5 minutes
  • Acrylic polymer crystals
  • Remains hydrated for up to 4 hours
  • Reusable
  • Available in a variety of colors and patterns like camo, stars and stripes, skulls, and flames

Specifications

Product Type Bandana
Color Red, White, Blue
For Use With (Application) Construction, Maintenance, Landscaping, Material Handling