missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6690 Cooling Arm Sleeves

Catalog No. 19819634
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19819634 Medium
19819635 Large
19819636 X-Large
19819637 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19819634 Supplier Ergodyne™ Supplier No. 12383

Chill-Its™ 6690 Cooling Arm Sleeves provide full arm coverage to absorb and wick away sweat, deliver advanced cooling relief, shield skin from UV rays, and even protect against light abrasions.

  • Performance knit fabric is soft, stretchable and durable
  • Instant cooling when wet that can last for hours
  • Wicks moisture when dry to absorb sweat from the body
  • UPF 50+ Protection
  • Protection from light abrasions
  • Reflective accents
  • Anti-odor treatment
  • Machine washable
  • Chemical-free cooling

Specifications

Material Performance Knit Fabric
Product Type Arm Sleeves
Size Medium
Color Black
For Use With (Application) Agriculture, Construction, Utilities, Transportation, Sports
Quantity 1/Ea.